Tisk…

5-Bromothiophene-3-carboxaldehyde 95%

BLDPBD164633-250MGEA 1140 CZK
BLDPBD164633-250MG BLDPBD164633-10G BLDPBD164633-5G BLDPBD164633-25G BLDPBD164633-1G
5-Bromothiophene-3-carboxaldehyde 95%
5-Bromothiophene-3-carboxaldehyde

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₅H₃BrOS
MW: 191,046 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09042555
CAS číslo: 18791-79-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR