Tisk…

1,5-Dibromo-2,4-difluorobenzene 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD164667-25GEA 1570 CZK
BLDPBD164667-25G BLDPBD164667-1G BLDPBD164667-5G BLDPBD164667-100G BLDPBD164667-10G
1,5-Dibromo-2,4-difluorobenzene 97%
1,5-Dibromo-2,4-difluorobenzene

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Vzorec: C₆H₂Br₂F₂
MW: 271,8849 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08458122
CAS číslo: 28342-75-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR