Tisk…

8-Methylene-1,4-dioxa-spiro[4.5]decane 98% + stabilizovaný

Dodavatel: BLD PHARMATECH GMBH


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

BLDPBD164723-1GEA 5900 CZK
BLDPBD164723-1G BLDPBD164723-250MG BLDPBD164723-5G
8-Methylene-1,4-dioxa-spiro[4.5]decane 98% + stabilizovaný
8-Methylene-1,4-dioxa-spiro[4.5]decane

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₉H₁₄O₂
MW: 154,206 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07779440
CAS číslo: 51656-90-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR