Tisk…

Methylterephthalonitrile 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD165165-5GEA 2710 CZK
BLDPBD165165-5G BLDPBD165165-25G BLDPBD165165-10G BLDPBD165165-250MG BLDPBD165165-1G
Methylterephthalonitrile 97%
Methylterephthalonitrile

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₉H₆N₂
MW: 142,157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00119249
CAS číslo: 55984-93-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR