Tisk…

Benzophenone-4-maleimide 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD16541-250MGEA 6790 CZK
BLDPBD16541-250MG BLDPBD16541-1G BLDPBD16541-100MG
Benzophenone-4-maleimide 97%
Benzophenone-4-maleimide

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₇H₁₁NO₃
MW: 277,2700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00079465
CAS číslo: 92944-71-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR