Tisk…

6-Quinolineboronic acid pinacol ester 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD165775-25GEA 16910 CZK
BLDPBD165775-25G BLDPBD165775-5G BLDPBD165775-1G BLDPBD165775-250MG
6-Quinolineboronic acid pinacol ester 98%
6-Quinolineboronic acid pinacol ester

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₅H₁₈BNO₂
MW: 255,1199 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06411327
CAS číslo: 406463-06-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR