Tisk…

5-(5-Chlorosulphonyl-2-ethoxyphenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD166933-25GEA 30900 CZK
BLDPBD166933-25G BLDPBD166933-1G BLDPBD166933-250MG BLDPBD166933-5G BLDPBD166933-10G
5-(5-Chlorosulphonyl-2-ethoxyphenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one 98%
5-(5-Chlorosulphonyl-2-ethoxyphenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₇H₁₉ClN₄O₄S
MW: 410,875 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09753597
CAS číslo: 139756-22-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR