Tisk…

1-(2,4,6-Triisopropylphenylsulphonyl)-1,2,4-triazole 98%

BLDPBD16767-10GEA 1770 CZK
BLDPBD16767-10G BLDPBD16767-25G BLDPBD16767-1G BLDPBD16767-5G
1-(2,4,6-Triisopropylphenylsulphonyl)-1,2,4-triazole 98%
1-(2,4,6-Triisopropylphenylsulphonyl)-1,2,4-triazole

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₇H₂₅N₃O₂S
MW: 335,47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014089
CAS číslo: 54230-60-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR