Tisk…

2-Methyl-2H-indazole-6-boronic acid pinacol ester 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD167811-250MGEA 1460 CZK
BLDPBD167811-250MG BLDPBD167811-1G
2-Methyl-2H-indazole-6-boronic acid pinacol ester 98%
2-Methyl-2H-indazole-6-boronic acid pinacol ester

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₄H₁₉BN₂O₂
MW: 258,1239 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11109435
CAS číslo: 1204580-79-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR