Tisk…

Isoquinoline-8-carbonitrile 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD167948-5GEA 10320 CZK
BLDPBD167948-5G BLDPBD167948-100MG BLDPBD167948-250MG BLDPBD167948-1G
Isoquinoline-8-carbonitrile 98%
Isoquinoline-8-carbonitrile

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₀H₆N₂
MW: 154,168 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10699232
CAS číslo: 362606-11-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR