Tisk…

3-Bromo-5-chloroanthranilic acid 95%

BLDPBD168272-250MGEA 656 CZK
BLDPBD168272-250MG BLDPBD168272-5G BLDPBD168272-25G BLDPBD168272-1G BLDPBD168272-100G
3-Bromo-5-chloroanthranilic acid 95%
3-Bromo-5-chloroanthranilic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₇H₅BrClNO₂
MW: 250,4771 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04035658
CAS číslo: 41198-02-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR