4-Fluoro-2-(trifluoromethyl)phenylboronic acid 98%
|
|
BLDPBD168784-25GEA
2740
CZK
BLDPBD168784-25G
BLDPBD168784-5G
BLDPBD168784-1G
BLDPBD168784-10G
4-Fluoro-2-(trifluoromethyl)phenylboronic acid 98%
4-Fluoro-2-(trifluoromethyl)phenylboronic acid
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Vzorec:
C₇H₅BF₄O₂ MW: 207,92 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08062379 CAS číslo: 182344-16-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...