Tisk…

Dicyclohexylphenylphosphine 98%

BLDPBD16960-100MGEA 542 CZK
BLDPBD16960-100MG BLDPBD16960-25G BLDPBD16960-250MG BLDPBD16960-1G BLDPBD16960-5G
Dicyclohexylphenylphosphine 98%
Dicyclohexylphenylphosphine

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₈H₂₇P
MW: 274,38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003854
CAS číslo: 6476-37-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR