Tisk…

2-(Boc-amino)-3-methylbutylamine 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD173829-250MGEA 1740 CZK
BLDPBD173829-250MG BLDPBD173829-1G BLDPBD173829-100MG
2-(Boc-amino)-3-methylbutylamine 98%
2-(Boc-amino)-3-methylbutylamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₀H₂₂N₂O₂
MW: 202,294 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12828716
CAS číslo: 871235-24-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR