Tisk…

2-Carbamoyl-6-nitrobenzoic acid 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD174264-5GEA 5850 CZK
BLDPBD174264-5G BLDPBD174264-250MG BLDPBD174264-1G
2-Carbamoyl-6-nitrobenzoic acid 95%
2-Carbamoyl-6-nitrobenzoic acid

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₈H₆N₂O₅
MW: 210,144 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00088952
CAS číslo: 65911-46-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR