Tisk…

2-Hydroxy-4'-(2-hydroxyethoxy)-2-methylpropiophenone 98%

BLDPBD17449-10GEA 171 CZK
BLDPBD17449-10G BLDPBD17449-500G BLDPBD17449-100G BLDPBD17449-25G
2-Hydroxy-4'-(2-hydroxyethoxy)-2-methylpropiophenone 98%
2-Hydroxy-4'-(2-hydroxyethoxy)-2-methylpropiophenone

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₁₂H₁₆O₄
MW: 224,2530 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00085267
CAS číslo: 106797-53-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR