Tisk…

Tetrakis(hydroxymethyl)phosphonium chloride ~80% in water

Dodavatel: BLD PHARMATECH GMBH

BLDPBD17854-25GEA 171 CZK
BLDPBD17854-25G
Tetrakis(hydroxymethyl)phosphonium chloride ~80% in water
Tetrakis(hydroxymethyl)phosphonium chloride

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₄H₁₂ClO₄P
MW: 190,56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00031687
CAS číslo: 124-64-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR