Tisk…

3-Amino-2-bromo-6-methoxypyridine 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD18146-25GEA 12660 CZK
BLDPBD18146-25G BLDPBD18146-10G BLDPBD18146-250MG BLDPBD18146-5G BLDPBD18146-1G BLDPBD18146-100G
3-Amino-2-bromo-6-methoxypyridine 98%
3-Amino-2-bromo-6-methoxypyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₆H₇BrN₂O
MW: 203,0400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05662394
CAS číslo: 135795-46-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR