Tisk…

2-Benzyloxy-5-chlorophenylboronic acid 96%

BLDPBD182531-5GEA 4620 CZK
BLDPBD182531-5G BLDPBD182531-25G BLDPBD182531-1G BLDPBD182531-10G BLDPBD182531-250MG
2-Benzyloxy-5-chlorophenylboronic acid 96%
2-Benzyloxy-5-chlorophenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₃H₁₂BClO₃
MW: 262,4966 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04039000
CAS číslo: 612832-83-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR