Tisk…

2-Aminoimidazole sulphate 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD18385-25GEA 1320 CZK
BLDPBD18385-25G BLDPBD18385-500G BLDPBD18385-100G BLDPBD18385-0.5G BLDPBD18385-5G
2-Aminoimidazole sulphate 97%
2-Aminoimidazole sulphate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₃H₇N₃O₄S
MW: 181,1700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00050811
CAS číslo: 36946-29-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR