Tisk…

2-Acrylamido-2-methylpropanesulphonic acid 98%

BLDPBD18525-25GEA 171 CZK
BLDPBD18525-25G
2-Acrylamido-2-methylpropanesulphonic acid 98%
2-Acrylamido-2-methylpropanesulphonic acid

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₇H₁₃NO₄S
MW: 207,25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00007522
CAS číslo: 15214-89-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR