Cyclopropanesulphonamide 98%
|
|
BLDPBD18563-1GEA
200
CZK
BLDPBD18563-1G
BLDPBD18563-100G
BLDPBD18563-0.5G
BLDPBD18563-500G
BLDPBD18563-25G
BLDPBD18563-5G
BLDPBD18563-10G
Cyclopropanesulphonamide 98%
Cyclopropanesulphonamide
Storage condition: Sealed in dry, Room Temperature
Vzorec:
C₃H₇NO₂S MW: 121,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08705286 CAS číslo: 154350-29-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...