Tisk…

2-Fluoro-4-nitrobenzaldehyde 98%

BLDPBD18621-1GEA 1230 CZK
BLDPBD18621-1G BLDPBD18621-5G BLDPBD18621-25G BLDPBD18621-10G BLDPBD18621-250MG
2-Fluoro-4-nitrobenzaldehyde 98%
2-Fluoro-4-nitrobenzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₇H₄FNO₃
MW: 169,10 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00968941
CAS číslo: 157701-72-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR