Tisk…

3-Carboxy-5-fluorophenylboronic acid 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD187790-5GEA 3940 CZK
BLDPBD187790-5G BLDPBD187790-1G BLDPBD187790-25G
3-Carboxy-5-fluorophenylboronic acid 98%
3-Carboxy-5-fluorophenylboronic acid

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₇H₆BFO₄
MW: 183,93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07363791
CAS číslo: 871329-84-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR