3-Quinolineboronic acid pinacol ester 98%
|
|
BLDPBD18840-5GEA
2260
CZK
BLDPBD18840-5G
BLDPBD18840-25G
BLDPBD18840-250MG
BLDPBD18840-1G
3-Quinolineboronic acid pinacol ester 98%
3-Quinolineboronic acid pinacol ester
Storage condition: Inert atmosphere, 2-8°C
Vzorec:
C₁₅H₁₈BNO₂ MW: 255,1199 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD05155218 CAS číslo: 171364-85-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...