2-Methoxy-4-methylaniline hydrochloride 98%
|
|
BLDPBD189468-5GEA
6360
CZK
BLDPBD189468-5G
BLDPBD189468-100MG
BLDPBD189468-1G
BLDPBD189468-250MG
2-Methoxy-4-methylaniline hydrochloride 98%
2-Methoxy-4-methylaniline hydrochloride
Storage condition: Inert atmosphere, Room Temperature
Vzorec:
C₈H₁₂ClNO MW: 173,6400 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09878311 CAS číslo: 71288-98-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...