Methyl-4-amino-3-methylbenzoate 97%
|
|
BLDPBD19019-10GEA
285
CZK
BLDPBD19019-10G
BLDPBD19019-500G
BLDPBD19019-100G
BLDPBD19019-5G
BLDPBD19019-1000G
BLDPBD19019-25G
Methyl-4-amino-3-methylbenzoate 97%
Methyl-4-amino-3-methylbenzoate
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Vzorec:
C₉H₁₁NO₂ MW: 165,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00102230 CAS číslo: 18595-14-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...