Methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate 95%
|
|
BLDPBD19060-1GEA
428
CZK
BLDPBD19060-1G
BLDPBD19060-25G
BLDPBD19060-5G
BLDPBD19060-0.025G
BLDPBD19060-250MG
Methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate 95%
Methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate
Storage condition: Sealed in dry, Room Temperature
Vzorec:
C₁₀H₉NO₃S MW: 223,25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00449104 CAS číslo: 188614-01-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...