Ethyl-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate 95+%
|
|
BLDPBD196691-1GEA
3480
CZK
BLDPBD196691-1G
BLDPBD196691-250MG
BLDPBD196691-5G
Ethyl-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate 95+%
Ethyl-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Storage condition: Inert atmosphere, Room Temperature
Vzorec:
C₁₁H₉F₃N₂O₂ MW: 258,20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11100212 CAS číslo: 860457-99-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...