Tisk…

2'-Chloro-4'-bromoacetophenone 97%

BLDPBD19698-5GEA 2880 CZK
BLDPBD19698-5G BLDPBD19698-25G BLDPBD19698-250MG BLDPBD19698-1G BLDPBD19698-10G
2'-Chloro-4'-bromoacetophenone 97%
2'-Chloro-4'-bromoacetophenone

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₈H₆BrClO
MW: 233,49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781036
CAS číslo: 252561-81-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR