Tisk…

5-Formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid 98%

BLDPBD19707-10GEA 656 CZK
BLDPBD19707-10G BLDPBD19707-1G BLDPBD19707-500G BLDPBD19707-100G BLDPBD19707-5G BLDPBD19707-25G
5-Formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid 98%
5-Formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C8H9NO3
MW: 167,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06202342
CAS číslo: 253870-02-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR