5-Chloro-2-(trifluoromethyl)benzyl alcohol 95%
|
|
BLDPBD19790-250MGEA
456
CZK
BLDPBD19790-250MG
BLDPBD19790-5G
BLDPBD19790-1G
5-Chloro-2-(trifluoromethyl)benzyl alcohol 95%
5-Chloro-2-(trifluoromethyl)benzyl alcohol
Storage condition: Sealed in dry, Room Temperature
Vzorec:
C₈H₆ClF₃O MW: 210,5800 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01631355 CAS číslo: 261763-21-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...