Tisk…

4-Formyl-1-naphthaleneboronic acid 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD20274-1GEA 15110 CZK
BLDPBD20274-1G BLDPBD20274-100MG BLDPBD20274-250MG
4-Formyl-1-naphthaleneboronic acid 98%
4-Formyl-1-naphthaleneboronic acid

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₁H₉BO₃
MW: 200,00 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01632205
CAS číslo: 332398-52-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR