Tisk…

2-Chloro-3-fluoro-6-methylpyridine 98%

BLDPBD20516-10GEA 7960 CZK
BLDPBD20516-10G BLDPBD20516-5G BLDPBD20516-25G BLDPBD20516-1G BLDPBD20516-250MG
2-Chloro-3-fluoro-6-methylpyridine 98%
2-Chloro-3-fluoro-6-methylpyridine

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₆H₅ClFN
MW: 145,56 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03095216
CAS číslo: 374633-32-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR