Tisk…

7-Methylindole-3-carboxyaldehyde 98%

BLDPBD20948-10GEA 4310 CZK
BLDPBD20948-10G BLDPBD20948-1G
7-Methylindole-3-carboxyaldehyde 98%
7-Methylindole-3-carboxyaldehyde

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Vzorec: C₁₀H₉NO
MW: 159,19 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00047170
CAS číslo: 4771-50-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR