5-Methylisophthalic acid 97%
|
|
BLDPBD21030-500GEA
9100
CZK
BLDPBD21030-500G
BLDPBD21030-10G
BLDPBD21030-25G
BLDPBD21030-100G
BLDPBD21030-5G
5-Methylisophthalic acid 97%
5-Methylisophthalic acid
Storage condition: Sealed in dry, Room Temperature
Vzorec:
C₉H₈O₄ MW: 180,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00013986 CAS číslo: 499-49-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...