Tisk…

Methyl-3-bromo-1H-indazole-5-carboxylate 97%

BLDPBD210997-1GEA 1370 CZK
BLDPBD210997-1G BLDPBD210997-250MG BLDPBD210997-5G BLDPBD210997-10G
Methyl-3-bromo-1H-indazole-5-carboxylate 97%
Methyl-3-bromo-1H-indazole-5-carboxylate

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₉H₇BrN₂O₂
MW: 255,0681 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11045401
CAS číslo: 1086391-06-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR