Tisk…

4-Bromo-3-fluoropyridine 98%

BLDPBD211993-1GEA 1940 CZK
BLDPBD211993-1G BLDPBD211993-25G
4-Bromo-3-fluoropyridine 98%
4-Bromo-3-fluoropyridine

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Vzorec: C₅H₃BrFN
MW: 175,9864 g/mol
Storage Temperature: Freezer
MDL Number: MFCD04113266
CAS číslo: 2546-52-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR