Tisk…

tert-Butyl 3,6-diazabicyclo[3.1.1]heptane-6-carboxylate 97%

BLDPBD212579-100MGEA 200 CZK
BLDPBD212579-100MG BLDPBD212579-1G BLDPBD212579-5G BLDPBD212579-10G BLDPBD212579-25G BLDPBD212579-250MG
tert-Butyl 3,6-diazabicyclo[3.1.1]heptane-6-carboxylate 97%
tert-Butyl 3,6-diazabicyclo[3.1.1]heptane-6-carboxylate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Vzorec: C₁₀H₁₈N₂O₂
MW: 198,262 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14581412
CAS číslo: 869494-16-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR