Tisk…

9-Bromo-10-(naphthalen-1-yl)anthracene 97%

BLDPBD212905-1GEA 285 CZK
BLDPBD212905-1G BLDPBD212905-25G BLDPBD212905-100G BLDPBD212905-5G
9-Bromo-10-(naphthalen-1-yl)anthracene 97%
9-Bromo-10-(naphthalen-1-yl)anthracene

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₂₄H₁₅Br
MW: 383,29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11046571
CAS číslo: 400607-04-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR