Tisk…

1,3-Dimethyl-1H-pyrazole-5-boronic acid pinacol ester 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD212940-1GEA 1230 CZK
BLDPBD212940-1G BLDPBD212940-100MG BLDPBD212940-25G BLDPBD212940-250MG BLDPBD212940-5G
1,3-Dimethyl-1H-pyrazole-5-boronic acid pinacol ester 97%
1,3-Dimethyl-1H-pyrazole-5-boronic acid pinacol ester

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₁H₁₉BN₂O₂
MW: 222,0918 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05664574
CAS číslo: 847818-79-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR