Tisk…

2-(Trifluoromethyl)-p-anisidine 98%

BLDPBD21304-25GEA 3140 CZK
BLDPBD21304-25G BLDPBD21304-5G BLDPBD21304-100G BLDPBD21304-10G BLDPBD21304-1G BLDPBD21304-0.025G
2-(Trifluoromethyl)-p-anisidine 98%
2-(Trifluoromethyl)-p-anisidine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₈H₈F₃NO
MW: 191,15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04972660
CAS číslo: 53903-49-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR