1,5-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester 95%
|
|
BLDPBD213101-100MGEA
656
CZK
BLDPBD213101-100MG
BLDPBD213101-250MG
BLDPBD213101-25G
BLDPBD213101-5G
BLDPBD213101-1G
1,5-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester 95%
1,5-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester
Storage condition: Inert atmosphere, Store in freezer, under -20°C
Vzorec:
C₁₁H₁₉BN₂O₂ MW: 222,0918 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD09864189 CAS číslo: 1036991-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...