Tisk…

6-Chloro-2-methyl-3H-imidazo[4,5-c]pyridine 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD213143-100MGEA 3880 CZK
BLDPBD213143-100MG BLDPBD213143-250MG
6-Chloro-2-methyl-3H-imidazo[4,5-c]pyridine 95%
6-Chloro-2-methyl-3H-imidazo[4,5-c]pyridine

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₇H₆ClN₃
MW: 167,596 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18256899
CAS číslo: 7205-42-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR