Tisk…

Methyl 1-methyl-4-nitro-1H-pyrrole-2-carboxylate 98%

BLDPBD214631-5GEA 1460 CZK
BLDPBD214631-5G BLDPBD214631-1G BLDPBD214631-25G
Methyl 1-methyl-4-nitro-1H-pyrrole-2-carboxylate 98%
Methyl 1-methyl-4-nitro-1H-pyrrole-2-carboxylate

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₇H₈N₂O₄
MW: 184,149 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03789112
CAS číslo: 13138-76-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR