(Trans-4-aminocyclohexyl)methanol 95%
|
|
BLDPBD215093-100MGEA
200
CZK
BLDPBD215093-100MG
BLDPBD215093-250MG
BLDPBD215093-5G
BLDPBD215093-1G
(Trans-4-aminocyclohexyl)methanol 95%
(Trans-4-aminocyclohexyl)methanol
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Vzorec:
C₇H₁₅NO MW: 129,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12407131 CAS číslo: 1467-84-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...