Tisk…

2,4-Dichloro-5-ethylpyrimidine 97%

BLDPBD215121-5GEA 10240 CZK
BLDPBD215121-5G BLDPBD215121-250MG BLDPBD215121-1G BLDPBD215121-25G BLDPBD215121-100MG BLDPBD215121-10G
2,4-Dichloro-5-ethylpyrimidine 97%
2,4-Dichloro-5-ethylpyrimidine

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₆H₆Cl₂N₂
MW: 177,031 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11040247
CAS číslo: 34171-40-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR