Tisk…

2,3-Difluoro-4-iodobenzaldehyde 97%

BLDPBD215201-250MGEA 2510 CZK
BLDPBD215201-250MG BLDPBD215201-100MG
2,3-Difluoro-4-iodobenzaldehyde 97%
2,3-Difluoro-4-iodobenzaldehyde

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₇H₃F₂IO
MW: 268,0000 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09835190
CAS číslo: 885590-99-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR