Tisk…

5-Bromo-2-chloroiodobenzene 95%

BLDPBD215210-100GEA 12400 CZK
BLDPBD215210-100G BLDPBD215210-5G BLDPBD215210-10G BLDPBD215210-25G BLDPBD215210-1G
5-Bromo-2-chloroiodobenzene 95%
5-Bromo-2-chloroiodobenzene

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Vzorec: C₆H₃BrClI
MW: 317,349 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07780669
CAS číslo: 774608-49-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR