Tisk…

3-Amino-2-chloroisonicotinic acid 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD216511-250MGEA 1860 CZK
BLDPBD216511-250MG BLDPBD216511-10G BLDPBD216511-1G BLDPBD216511-25G BLDPBD216511-5G
3-Amino-2-chloroisonicotinic acid 97%
3-Amino-2-chloroisonicotinic acid

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₆H₅ClN₂O₂
MW: 172,569 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15143308
CAS číslo: 58483-94-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR